(5-Nitro-1-benzofuran-2-yl)methanol ≥95%
Supplier: Matrix Scientific
046458-500MG
046458-1G
046458-5G
101875-574EA
375.38
USD
101875-574
101875-576
101875-578
(5-Nitro-1-benzofuran-2-yl)methanol ≥95%
(5-Nitro-1-benzofuran-2-yl)methanol
Hazard: Irritant
Formula:
C₉H₇NO₄ Melting Pt: 117…119 °C |
MDL Number:
MFCD10758066 CAS Number: 90322-48-8 |
Specification Test Results
Melting point | 117-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...