3-Nitro-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
046456-500MG
046456-1G
101875-570EA
556.92
USD
101875-570
101875-572
3-Nitro-1H-pyrrolo[2,3-b]pyridine ≥95%
3-Nitro-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₇H₅N₃O₂ MW: 163.14 g/mol Melting Pt: 309…311 °C |
MDL Number:
MFCD08272225 CAS Number: 23709-47-9 |
Specification Test Results
Melting point | 309-311°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...