Methyl-4-iodo-1H-pyrrole-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-iodopyrrole-2-carboxylate
046452-500MG
046452-1G
046452-5G
101875-554EA
73.85
USD
101875-554
101875-556
101875-558
Methyl-4-iodo-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-iodo-1H-pyrrole-2-carboxylate
Hazard: Irritant
Formula:
C₆H₆INO₂ MW: 251.023 g/mol Melting Pt: 77…78 °C |
MDL Number:
MFCD07368824 CAS Number: 40740-41-8 |
Specification Test Results
Melting point | 77-78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...