4-Iodo-2-(trichloroacetyl)pyrrole ≥95%
Supplier: Matrix Scientific
046451-500MG
046451-1G
046451-5G
101875-548EA
375.38
USD
101875-548
101875-550
101875-552
4-Iodo-2-(trichloroacetyl)pyrrole ≥95%
4-Iodo-2-(trichloroacetyl)pyrrole
Hazard: Irritant
Formula:
C₆H₃Cl₃INO Melting Pt: 143…144 °C |
MDL Number:
MFCD08277257 CAS Number: 72652-33-6 |
Specification Test Results
Melting point | 143-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...