4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

046450-500MG 046450-1G 046450-5G
101875-542EA 375.38 USD
101875-542 101875-544 101875-546
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈F₃NO
MW: 251.21 g/mol
Melting Pt: 67…68 °C
MDL Number: MFCD10758064
CAS Number: 847446-85-9

Order Now

Specification Test Results

Melting point 67-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR