5-Bromobenzoxazole ≥95%

Supplier: Matrix Scientific

046440-500MG 046440-1G 046440-5G
101875-504EA 21.54 USD
101875-504 101875-506 101875-508
5-Bromobenzoxazole ≥95%
5-Bromobenzoxazole

Hazard: Irritant

Formula: C₇H₄BrNO
Melting Pt: 38-40°C
MDL Number: MFCD03095026
CAS Number: 132244-31-6

Order Now

Specification Test Results

Melting point 38-40°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR