Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

046439-500MG 046439-1G 046439-5G
101875-498EA 278.46 USD
101875-498 101875-500 101875-502
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈Cl₂N₂O₂
MW: 259.089 g/mol
Melting Pt: 169…170 °C
MDL Number: MFCD02325395
CAS Number: 1431965-03-5

Order Now

Specification Test Results

Melting point 169-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR