Ethyl-3-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Iodo-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester, ethyl 3-iodoimidazo[1,2-a]pyridine-2-carboxylate
046437-500MG
046437-1G
101875-488EA
320
USD
101875-488
101875-490
Ethyl-3-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-iodoimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉IN₂O₂ Melting Pt: 144-146°C |
MDL Number:
MFCD06739188 CAS Number: 292858-07-2 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...