3'-Nitro[1,1'-biphenyl]-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
046433-500MG
046433-1G
046433-5G
101875-470EA
278.46
USD
101875-470
101875-472
101875-474
3'-Nitro[1,1'-biphenyl]-4-carboxylic acid ≥95%
3'-Nitro[1,1'-biphenyl]-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₉NO₄ Melting Pt: >300 °C |
MDL Number:
MFCD01846432 CAS Number: 5737-85-9 |
Specification Test Results
Melting point | >300°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...