N-{2,4-Dichloro-5-[(4-nitrobenzyl)oxy]phenyl}acetamide ≥95%

Supplier: Matrix Scientific

046431-500MG 046431-1G 046431-5G
101875-462EA 375.38 USD
101875-462 101875-464 101875-466
N-{2,4-Dichloro-5-[(4-nitrobenzyl)oxy]phenyl}acetamide ≥95%
N-{2,4-Dichloro-5-[(4-nitrobenzyl)oxy]phenyl}acetamide

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂N₂O₄
MW: 355.18 g/mol
Melting Pt: 189…190 °C
MDL Number: MFCD10758057
CAS Number: 1033463-19-2

Order Now

Specification Test Results

Melting point 189-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR