Methyl-4-bromo-5-formyl-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromo-5-formylthiophene-2-carboxylate
046427-500MG
046427-1G
101875-448EA
747.69
USD
101875-448
101875-450
Methyl-4-bromo-5-formyl-2-thiophenecarboxylate ≥95%
Methyl-4-bromo-5-formyl-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₇H₅BrO₃S Melting Pt: 95…97 °C |
MDL Number:
MFCD11100203 CAS Number: 648412-59-3 |
Specification Test Results
Melting point | 95-97°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...