2-Amino-6-bromo-4-quinolinol ≥95%
Supplier: Matrix Scientific
046426-500MG
046426-1G
101875-444EA
210.77
USD
101875-444
101875-446
2-Amino-6-bromo-4-quinolinol ≥95%
2-Amino-6-bromo-4-quinolinol
Hazard: Irritant
Formula:
C₉H₇BrN₂O MW: 239.08 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD08234781 CAS Number: 123420-09-7 |
Specification Test Results
Melting point | >300°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...