1-Methyl-6-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

046422-500MG 046422-1G
101875-426EA 556.92 USD
101875-426 101875-428
1-Methyl-6-nitro-1H-indazole ≥95%
1-Methyl-6-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
MW: 177.160 g/mol
Melting Pt: 122-123°C
MDL Number: MFCD00456124
CAS Number: 6850-23-3

Order Now

Specification Test Results

Melting point 122-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR