N-[4-(2-Amino-1,3-thiazol-4-yl)phenyl]acetamide ≥95%
Supplier: Matrix Scientific
046416-500MG
046416-1G
046416-5G
101875-408EA
375.38
USD
101875-408
101875-410
101875-412
N-[4-(2-Amino-1,3-thiazol-4-yl)phenyl]acetamide ≥95%
N-[4-(2-Amino-1,3-thiazol-4-yl)phenyl]acetamide
Hazard: Irritant
Formula:
C₁₁H₁₁N₃OS MW: 233.29 g/mol Melting Pt: 251…212 °C |
MDL Number:
MFCD00207103 CAS Number: 21674-96-4 |
Specification Test Results
Melting point | 251-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...