Methyl amino(3-fluorophenyl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

046412-500MG 046412-1G
101875-388EA 375.38 USD
101875-388 101875-390
Methyl amino(3-fluorophenyl)acetate hydrochloride ≥95%
Methyl amino(3-fluorophenyl)acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFNO₂
MW: 219.64 g/mol
Melting Pt: 21 °C
MDL Number: MFCD09152736
CAS Number: 42718-21-8

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR