Methyl amino(4-methoxyphenyl)acetate hydrochloride ≥95%
Supplier: Matrix Scientific
046411-500MG
046411-1G
101875-384EA
553.85
USD
101875-384
101875-386
Methyl amino(4-methoxyphenyl)acetate hydrochloride ≥95%
Methyl amino(4-methoxyphenyl)acetate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClNO₃ Melting Pt: 183…186 °C |
MDL Number:
MFCD09152735 CAS Number: 74273-47-5 |
Specification Test Results
Melting point | 183-186°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...