Methyl amino(4-methylphenyl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-2-(p-tolyl)acetate hydrochloride

046410-500MG 046410-1G
101875-380EA 375.38 USD
101875-380 101875-382
Methyl amino(4-methylphenyl)acetate hydrochloride ≥95%
Methyl amino(4-methylphenyl)acetate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Melting Pt: 193…195 °C
MDL Number: MFCD08456676
CAS Number: 134722-09-1

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR