Methyl amino(4-fluorophenyl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-DL-phenylglycine methyl ester hydrochloride, Methyl 2-amino-2-(4-fluorophenyl)acetate hydrochloride

046408-500MG 046408-1G
101875-372EA 118.46 USD
101875-372 101875-374
Methyl amino(4-fluorophenyl)acetate hydrochloride ≥95%
Methyl amino(4-fluorophenyl)acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFNO₂
MW: 219.64 g/mol
Melting Pt: 177…179 °C
MDL Number: MFCD09152734
CAS Number: 42718-18-3

Order Now

Specification Test Results

Melting point 177-179°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR