3-Chloro-2-pyridinethiol ≥95%

Supplier: Matrix Scientific

046404-500MG 046404-1G
101875-354EA 375.38 USD
101875-354 101875-356
3-Chloro-2-pyridinethiol ≥95%
3-Chloro-2-pyridinethiol

Hazard: Irritant

Formula: C₅H₄ClNS
Melting Pt: 198…200 °C
MDL Number: MFCD00661364
CAS Number: 5897-94-9

Order Now

Specification Test Results

Melting point 198-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR