2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

046403-500MG 046403-1G 046403-5G
101875-348EA 400 USD
101875-348 101875-350 101875-352
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylic acid ≥95%
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₉H₆ClN₃O₂
Melting Pt: 242…244 °C
MDL Number: MFCD09152733
CAS Number: 220141-15-1

Order Now

Specification Test Results

Melting point 242-244°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR