3-Nitro-2-pyridinethiol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Nitropyridine-2(1H)-thione, 3-Nitropyridine-2-thiol

046402-500MG 046402-1G
101875-344EA 160 USD
101875-344 101875-346
3-Nitro-2-pyridinethiol ≥95%
3-Nitro-2-pyridinethiol

Hazard: Irritant

Formula: C₅H₄N₂O₂S
MW: 156.16 g/mol
Melting Pt: 173…174 °C
MDL Number: MFCD00661369
CAS Number: 38240-29-8

Order Now

Specification Test Results

Melting point 173-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR