4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
046401-500MG
046401-1G
101875-340EA
556.92
USD
101875-340
101875-342
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbohydrazide ≥95%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbohydrazide
Hazard: Irritant
Formula:
C₁₆H₁₆N₂O₃ Melting Pt: 160…162 °C |
MDL Number:
MFCD09152732 CAS Number: 952183-14-1 |
Specification Test Results
Melting point | 160-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...