4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzoic acid
046400-500MG
046400-1G
101875-336EA
375.38
USD
101875-336
101875-338
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid ≥95%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₄ MW: 270.29 g/mol Melting Pt: 217…220 °C |
MDL Number:
MFCD09152731 CAS Number: 952183-13-0 |
Specification Test Results
Melting point | 217-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...