2-Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoic acid ≥95%

Supplier: Matrix Scientific

046399-500MG 046399-1G
101875-332EA 556.92 USD
101875-332 101875-334
2-Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoic acid ≥95%
2-Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₄S
Melting Pt: 60…62 °C
MDL Number: MFCD09607896
CAS Number: 952183-12-9

Order Now

Specification Test Results

Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR