2-Chloro-4-piperidinobenzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
046395-500MG
046395-1G
046395-5G
101875-320EA
375.38
USD
101875-320
101875-322
101875-324
2-Chloro-4-piperidinobenzenecarboxylic acid ≥95%
2-Chloro-4-piperidinobenzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₄ClNO₂ Melting Pt: 224…226 °C |
MDL Number:
MFCD09152730 CAS Number: 313674-11-2 |
Specification Test Results
Melting point | 224-226°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...