2-Chloro-N-(2-hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
046394-500MG
046394-1G
046394-5G
101875-314EA
375.38
USD
101875-314
101875-316
101875-318
2-Chloro-N-(2-hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide ≥95%
2-Chloro-N-(2-hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₃O₂ Melting Pt: 168…170 °C |
MDL Number:
MFCD09152729 CAS Number: 952183-10-7 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...