Ethyl-4-(4-phenoxybenzoyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

046393-500MG 046393-1G 046393-5G
101875-308EA 375.38 USD
101875-308 101875-310 101875-312
Ethyl-4-(4-phenoxybenzoyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(4-phenoxybenzoyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₉H₁₅NO₅
Melting Pt: 63…65 °C
MDL Number: MFCD09152728
CAS Number: 952183-09-4

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR