2-(4-Nitrophenyl)-2-oxoethyl acetate ≥95%

Supplier: Matrix Scientific

046390-500MG 046390-1G 046390-5G
101875-290EA 161.54 USD
101875-290 101875-292 101875-294
2-(4-Nitrophenyl)-2-oxoethyl acetate ≥95%
2-(4-Nitrophenyl)-2-oxoethyl acetate

Hazard: Irritant

Formula: C₁₀H₉NO₅
Melting Pt: 122…124 °C
MDL Number: MFCD09152776
CAS Number: 65921-30-4

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR