2-(4-Methoxyphenyl)-2-oxoethyl acetate ≥95%
Supplier: Matrix Scientific
046389-500MG
046389-1G
046389-5G
101875-284EA
176.92
USD
101875-284
101875-286
101875-288
2-(4-Methoxyphenyl)-2-oxoethyl acetate ≥95%
2-(4-Methoxyphenyl)-2-oxoethyl acetate
Hazard: Irritant
Formula:
C₁₁H₁₂O₄ Melting Pt: 63…65 °C |
MDL Number:
MFCD00540316 CAS Number: 58518-78-8 |
Specification Test Results
Melting point | 63-65°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...