4-Cyano-2-pyridinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Cyanopicolinic acid
046384-500MG
046384-1G
101875-260EA
692.31
USD
101875-260
101875-262
4-Cyano-2-pyridinecarboxylic acid ≥95%
4-Cyano-2-pyridinecarboxylic acid
Hazard: Irritant
Formula:
C₇H₄N₂O₂ MW: 148.121 g/mol Melting Pt: 210 °C (dec.) |
MDL Number:
MFCD09152773 CAS Number: 640296-19-1 |
Specification Test Results
Melting point | 210°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...