Ethyl-5-chloro-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloropicolinate

046382-500MG 046382-1G
101875-256EA 181.54 USD
101875-256 101875-258
Ethyl-5-chloro-2-pyridinecarboxylate ≥95%
Ethyl-5-chloro-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 47…50 °C
MDL Number: MFCD07772018
CAS Number: 128072-93-5

Order Now

Specification Test Results

Melting point 47-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR