Ethyl-5-cyano-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

046381-500MG 046381-1G
101875-252EA 461.54 USD
101875-252 101875-254
Ethyl-5-cyano-2-pyridinecarboxylate ≥95%
Ethyl-5-cyano-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Melting Pt: 74…76 °C
MDL Number: MFCD09152772
CAS Number: 41051-03-0

Order Now

Specification Test Results

Melting point 74-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR