Ethyl-4-cyano-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

046378-500MG 046378-1G
101875-238EA 618.46 USD
101875-238 101875-240
Ethyl-4-cyano-2-pyridinecarboxylate ≥95%
Ethyl-4-cyano-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
Melting Pt: 91…93 °C
MDL Number: MFCD09152770
CAS Number: 97316-50-2

Order Now

Specification Test Results

Melting point 91-93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR