6-(Ethoxycarbonyl)nicotinic acid ≥95%

Supplier: Matrix Scientific

046377-500MG 046377-1G
101875-234EA 260 USD
101875-234 101875-236
6-(Ethoxycarbonyl)nicotinic acid ≥95%
6-(Ethoxycarbonyl)nicotinic acid

Hazard: Irritant

Formula: C₉H₉NO₄
Melting Pt: >240 °C
MDL Number: MFCD09607895
CAS Number: 17874-78-1

Order Now

Specification Test Results

Melting point >240°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR