Ethyl-3-cyano-2-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
046376-500MG
046376-1G
101875-230EA
921.54
USD
101875-230
101875-232
Ethyl-3-cyano-2-pyridinecarboxylate ≥95%
Ethyl-3-cyano-2-pyridinecarboxylate
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol Melting Pt: 93…95 °C |
MDL Number:
MFCD07368151 CAS Number: 97316-55-7 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...