5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Dihydro-5-[(4-methylpiperazin-1-yl)methyl]-2-thioxopyrimidin-4(1H)-one
046371-500MG
046371-1G
046371-5G
101875-212EA
181.54
USD
101875-212
101875-214
101875-216
5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%
5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Hazard: Irritant
Formula:
C₁₀H₁₆N₄OS MW: 240.33 g/mol Melting Pt: 227…228 °C |
MDL Number:
MFCD09607894 CAS Number: 952183-05-0 |
Specification Test Results
Melting point | 227-228°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...