5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-5-[(morpholin-4-yl)methyl]-2-thioxopyrimidin-4(1H)-one

046369-500MG 046369-1G 046369-5G
101875-200EA 181.54 USD
101875-200 101875-202 101875-204
5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%
5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone

Hazard: Irritant

Formula: C₉H₁₃N₃O₂S
MW: 227.29 g/mol
Melting Pt: 223…224 °C
MDL Number: MFCD01689081
CAS Number: 89665-74-7

Order Now

Specification Test Results

Melting point 223-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR