Benzyl-N-[2-methyl-1-(phenylsulfonyl)propyl]carbamate ≥95%

Supplier: Matrix Scientific

046368-500MG 046368-1G 046368-5G
101875-194EA 181.54 USD
101875-194 101875-196 101875-198
Benzyl-N-[2-methyl-1-(phenylsulfonyl)propyl]carbamate ≥95%
Benzyl-N-[2-methyl-1-(phenylsulfonyl)propyl]carbamate

Hazard: Irritant

Formula: C₁₈H₂₁NO₄S
MW: 347.44 g/mol
Melting Pt: 100…101 °C
MDL Number: MFCD03012203
CAS Number: 439095-25-7

Order Now

Specification Test Results

Melting point 100-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR