N-(3-Bromophenyl)-N'-(2-chloroethyl)urea ≥95%

Supplier: Matrix Scientific

046362-500MG 046362-1G 046362-5G
101875-166EA 278.46 USD
101875-166 101875-168 101875-170
N-(3-Bromophenyl)-N'-(2-chloroethyl)urea ≥95%
N-(3-Bromophenyl)-N'-(2-chloroethyl)urea

Hazard: Irritant

Formula: C₉H₁₀BrClN₂O
Melting Pt: 108…110 °C
MDL Number: MFCD00451942
CAS Number: 146257-17-2

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR