Ethyl-1-(4-methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3,5-dimethyl-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylate
046359-500MG
046359-1G
046359-5G
101875-148EA
181.54
USD
101875-148
101875-150
101875-152
Ethyl-1-(4-methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(4-methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₈N₂O₃ MW: 274.32 g/mol Melting Pt: 50…52 °C |
MDL Number:
MFCD08443956 CAS Number: 849416-70-2 |
Specification Test Results
Melting point | 50-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...