5-Amino-3-(3-bromophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

046356-500MG 046356-1G 046356-5G
101875-136EA 78.46 USD
101875-136 101875-138 101875-140
5-Amino-3-(3-bromophenyl)-1H-pyrazole ≥95%
5-Amino-3-(3-bromophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₈BrN₃
Melting Pt: 149-151°C
MDL Number: MFCD06738756
CAS Number: 149246-81-1

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR