5-Amino-3-(3-bromophenyl)isoxazole ≥95%

Supplier: Matrix Scientific

046355-500MG 046355-1G 046355-5G
101875-130EA 200 USD
101875-130 101875-132 101875-134
5-Amino-3-(3-bromophenyl)isoxazole ≥95%
5-Amino-3-(3-bromophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.0687 g/mol
Melting Pt: 119-121°C
MDL Number: MFCD07787076
CAS Number: 119162-52-6

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR