3-(3,5-Dimethoxyphenyl)-5-isoxazolamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-(3,5-dimethoxyphenyl)isoxazole

046354-500MG 046354-1G 046354-5G
101875-124EA 278.46 USD
101875-124 101875-126 101875-128
3-(3,5-Dimethoxyphenyl)-5-isoxazolamine ≥95%
3-(3,5-Dimethoxyphenyl)-5-isoxazolamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD08443955
CAS Number: 924868-82-6

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR