3-(3,4-Dimethoxyphenyl)-5-isoxazolamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-(3,4-dimethoxyphenyl)isoxazole

046351-500MG 046351-1G 046351-5G
101875-106EA 278.46 USD
101875-106 101875-108 101875-110
3-(3,4-Dimethoxyphenyl)-5-isoxazolamine ≥95%
3-(3,4-Dimethoxyphenyl)-5-isoxazolamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 159…161 °C
MDL Number: MFCD07382987
CAS Number: 501325-88-8

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR