3-(3,5-Dimethoxyphenyl)-3-oxopropanenitrile ≥95%
Supplier: Matrix Scientific
046349-500MG
046349-1G
046349-5G
101875-100EA
181.54
USD
101875-100
101875-102
101875-104
3-(3,5-Dimethoxyphenyl)-3-oxopropanenitrile ≥95%
3-(3,5-Dimethoxyphenyl)-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₁H₁₁NO₃ Melting Pt: 121…123 °C |
MDL Number:
MFCD00158877 CAS Number: 70988-04-4 |
Specification Test Results
Melting point | 121-123°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...