tert-Butyl-2-(2-cyanophenoxy)acetate ≥95%

Supplier: Matrix Scientific

046345-500MG 046345-1G 046345-5G
101875-082EA 181.54 USD
101875-082 101875-084 101875-086
tert-Butyl-2-(2-cyanophenoxy)acetate ≥95%
tert-Butyl-2-(2-cyanophenoxy)acetate

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Melting Pt: 44…45 °C
MDL Number: MFCD09972227
CAS Number: 609804-44-6

Order Now

Specification Test Results

Melting point 44-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR