5-Chloro-2-methyl-6-(1-pyrrolidinyl)-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
046279-500MG
046279-1G
101874-924EA
556.92
USD
101874-924
101874-926
5-Chloro-2-methyl-6-(1-pyrrolidinyl)-4-pyrimidinol ≥95%
5-Chloro-2-methyl-6-(1-pyrrolidinyl)-4-pyrimidinol
Hazard: Irritant
Formula:
C₉H₁₂ClN₃O Melting Pt: 261…265 °C |
MDL Number:
MFCD12025849 CAS Number: 1135283-15-6 |
Specification Test Results
Melting point | 261-265°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...