5-Chloro-2-methoxy-nicotinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloro-2-methoxypyridine-3-carboxylic acid, 5-Chloro-2-methoxynicotinic acid
046278-1G
046278-5G
101874-922EA
16.92
USD
101874-922
102551-346
5-Chloro-2-methoxy-nicotinic acid ≥95%
5-Chloro-2-methoxy-nicotinic acid
Hazard: Irritant
Formula:
C₇H₆ClNO₃ Melting Pt: 146-148°C |
MDL Number:
MFCD08236790 CAS Number: 54916-65-3 |
Specification Test Results
Melting point | 146-148°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...