Ethyl-1-{[(tert-butoxycarbonyl)(2-chloroethyl)amino]sulfonyl}-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

046277-500MG 046277-1G
101874-916EA 375.38 USD
101874-916 101874-918
Ethyl-1-{[(tert-butoxycarbonyl)(2-chloroethyl)amino]sulfonyl}-4-piperidinecarboxylate ≥95%
Ethyl-1-{[(tert-butoxycarbonyl)(2-chloroethyl)amino]sulfonyl}-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₇ClN₂O₆S
Melting Pt: 82…83 °C
MDL Number: MFCD12025848
CAS Number: 1135283-14-5

Order Now

Specification Test Results

Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR