1-Ethynyl-2-methoxy-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
046274-500MG
046274-1G
046274-5G
101874-908EA
556.92
USD
101874-908
101874-910
101874-912
1-Ethynyl-2-methoxy-4-nitrobenzene ≥95%
1-Ethynyl-2-methoxy-4-nitrobenzene
Hazard: Irritant
Formula:
C₉H₇NO₃ Melting Pt: 130…131 °C |
MDL Number:
MFCD11840985 CAS Number: 1135283-10-1 |
Specification Test Results
Melting point | 130-131°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...